Assay Details
| Gene Symbol: mas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111858.1 | NP_001105328.1 | 1108 | 93 | ||
| GenBank mRNA | AY108138.2 | - | 1214 | 93 | ||
| L35914.1 | - | 1108 | 93 | |||
More Information
Set Membership: |
|

mas1
Genomic Map