Assay Details
| Gene Symbol: an1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111859.1 | NP_001105329.1 | 1738 | 67 | ||
| GenBank mRNA | AY109636.1 | - | 1738 | 67 | ||
| L37750.1 | - | 1738 | 67 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

an1| Gene Symbol: an1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111859.1 | NP_001105329.1 | 1738 | 67 | ||
| GenBank mRNA | AY109636.1 | - | 1738 | 67 | ||
| L37750.1 | - | 1738 | 67 | |||