Assay Details
| Gene Symbol: pal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111864.1 | NP_001105334.1 | 480 | 76 | ||
| GenBank mRNA | L77912.1 | - | 480 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pal3| Gene Symbol: pal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111864.1 | NP_001105334.1 | 480 | 76 | ||
| GenBank mRNA | L77912.1 | - | 480 | 76 | ||
Set Membership: |
|