Assay Details
| Gene Symbol: phi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111898.1 | NP_001105368.1 | 797 | 99 | ||
| GenBank mRNA | BT042918.1 | - | 927 | 99 | ||
| BT065011.1 | - | 1090 | 99 | |||
| EU963540.1 | - | 863 | 99 | |||
| U17225.1 | - | 797 | 99 | |||
More Information
Set Membership: |
|

phi1
Genomic Map