Assay Details
| Gene Symbol: eno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111901.1 | NP_001105371.1 | 93 | 68 | ||
| GenBank mRNA | AY103866.1 | - | 230 | 68 | ||
| U17973.1 | - | 118 | 68 | |||
More Information
Set Membership: |
|

eno2
Genomic Map