Assay Details
| Gene Symbol: adss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111916.1 | NP_001105386.1 | 297 | 76 | ||
| GenBank mRNA | U49388.1 | - | 297 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

adss1| Gene Symbol: adss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111916.1 | NP_001105386.1 | 297 | 76 | ||
| GenBank mRNA | U49388.1 | - | 297 | 76 | ||
Set Membership: |
|