Assay Details
| Gene Symbol: cat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111945.1 | NP_001105415.1 | 283 | 98 | ||
| GenBank mRNA | X12538.1 | - | 283 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cat1| Gene Symbol: cat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111945.1 | NP_001105415.1 | 283 | 98 | ||
| GenBank mRNA | X12538.1 | - | 283 | 98 | ||
Set Membership: |
|