Assay Details
| Gene Symbol: pdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111952.1 | NP_001105422.1 | 1695 | 107 | ||
| GenBank mRNA | AY109914.1 | - | 107 | |||
| X17555.1 | - | 1695 | 107 | |||
More Information
Set Membership: |
|

pdc1
Genomic Map