Assay Details
| Gene Symbol: ptk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111954.1 | NP_001105424.1 | 2351 | 83 | ||
| GenBank mRNA | AY111028.1 | - | 83 | |||
| X52384.1 | - | 2351 | 83 | |||
More Information
Set Membership: |
|

ptk1
Genomic Map