Assay Details
| Gene Symbol: TIDP2849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111956.1 | NP_001105426.1 | 759 | 65 | ||
| GenBank mRNA | X52879.1 | - | 759 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIDP2849| Gene Symbol: TIDP2849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111956.1 | NP_001105426.1 | 759 | 65 | ||
| GenBank mRNA | X52879.1 | - | 759 | 65 | ||
Set Membership: |
|