Assay Details
| Gene Symbol: gln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111973.1 | NP_001105443.1 | 820 | 60 | ||
| GenBank mRNA | AY104270.1 | - | 820 | 60 | ||
| X65927.1 | - | 820 | 60 | |||
More Information
Set Membership: |
|

gln2
Genomic Map