Assay Details
| Gene Symbol: pme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112017.1 | NP_001105487.1 | 997 | 71 | ||
| GenBank mRNA | AY109517.1 | - | 997 | 71 | ||
| Y13285.1 | - | 997 | 71 | |||
More Information
Set Membership: |
|

pme1
Genomic Map