Assay Details
| Gene Symbol: dnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112042.1 | NP_001105512.1 | 1006 | 101 | ||
| GenBank mRNA | AF288075.1 | - | 1006 | 101 | ||
| AY111987.1 | - | 101 | ||||
More Information
Set Membership: |
|

dnp1
Genomic Map