Assay Details
| Gene Symbol: mlo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112057.1 | NP_001105527.1 | 600 | 98 | ||
| GenBank mRNA | AY029314.1 | - | 600 | 98 | ||
| AY108219.1 | - | 584 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlo3| Gene Symbol: mlo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112057.1 | NP_001105527.1 | 600 | 98 | ||
| GenBank mRNA | AY029314.1 | - | 600 | 98 | ||
| AY108219.1 | - | 584 | 98 | |||