Assay Details
Gene Symbol: mlo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112057.1 | NP_001105527.1 | 910 | 78 | ||
GenBank mRNA | AY029314.1 | - | 910 | 78 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: mlo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112057.1 | NP_001105527.1 | 910 | 78 | ||
GenBank mRNA | AY029314.1 | - | 910 | 78 |