Assay Details
| Gene Symbol: pco103506(726) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112100.1 | NP_001105570.1 | 1398 | 77 | ||
| GenBank mRNA | AB112936.1 | - | 1398 | 77 | ||
More Information
Set Membership: |
|

pco103506(726)
Genomic Map