Assay Details
| Gene Symbol: mad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112148.1 | NP_001105618.1 | 438 | 98 | ||
| GenBank mRNA | AF143681.1 | - | 464 | 98 | ||
| BT069597.1 | - | 538 | 98 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

mad2| Gene Symbol: mad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112148.1 | NP_001105618.1 | 438 | 98 | ||
| GenBank mRNA | AF143681.1 | - | 464 | 98 | ||
| BT069597.1 | - | 538 | 98 | |||