Assay Details
| Gene Symbol: mad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112148.1 | NP_001105618.1 | 784 | 67 | ||
| GenBank mRNA | AF143681.1 | - | 810 | 67 | ||
| BT069597.1 | - | 884 | 67 | |||
| EU975638.1 | - | 802 | 67 | |||
More Information
Set Membership: |
|

mad2
Genomic Map