Assay Details
| Gene Symbol: hdt102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112161.1 | NP_001105631.1 | 852 | 59 | ||
| GenBank mRNA | AF254072.1 | - | 852 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hdt102| Gene Symbol: hdt102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112161.1 | NP_001105631.1 | 852 | 59 | ||
| GenBank mRNA | AF254072.1 | - | 852 | 59 | ||
Set Membership: |
|