Assay Details
| Gene Symbol: pk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112185.1 | NP_001105655.1 | 2350 | 93 | ||
| GenBank mRNA | AJ010166.1 | - | 2350 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pk3| Gene Symbol: pk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112185.1 | NP_001105655.1 | 2350 | 93 | ||
| GenBank mRNA | AJ010166.1 | - | 2350 | 93 | ||
Set Membership: |
|