Assay Details
| Gene Symbol: mlo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112190.1 | NP_001105660.1 | 978 | 58 | ||
| GenBank mRNA | AY029312.2 | - | 978 | 58 | ||
| BT036101.1 | - | 1083 | 58 | |||
More Information
Set Membership: |
|

mlo1
Genomic Map