Assay Details
| Gene Symbol: mstr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112211.1 | NP_001105681.1 | 486 | 95 | ||
| GenBank mRNA | AY683004.1 | - | 486 | 95 | ||
| EU956633.1 | - | 569 | 95 | |||
More Information
Set Membership: |
|

mstr1
Genomic Map