Assay Details
| Gene Symbol: gln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112255.1 | NP_001105725.1 | 1121 | 76 | ||
| GenBank mRNA | AY110796.1 | - | 1121 | 76 | ||
| X65931.1 | - | 1121 | 76 | |||
More Information
Set Membership: |
|

gln1
Genomic Map