Assay Details
| Gene Symbol: eif3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112256.1 | NP_001105726.1 | 1296 | 87 | ||
| GenBank mRNA | AF073329.1 | - | 1296 | 87 | ||
| AY109549.1 | - | 1296 | 87 | |||
More Information
Set Membership: |
|

eif3
Genomic Map