Assay Details
| Gene Symbol: TPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112263.1 | NP_001105733.1 | 1119 | 100 | ||
| GenBank mRNA | AY518312.1 | - | 1119 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPS4| Gene Symbol: TPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112263.1 | NP_001105733.1 | 1119 | 100 | ||
| GenBank mRNA | AY518312.1 | - | 1119 | 100 | ||