Assay Details
| Gene Symbol: pdi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112286.1 | NP_001105756.1 | 1449 | 105 | ||
| GenBank mRNA | AY739286.1 | - | 1449 | 105 | ||
| BT087465.1 | - | 1446 | 105 | |||
More Information
Set Membership: |
|

pdi3
Genomic Map