Assay Details
| Gene Symbol: pdi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112287.1 | NP_001105757.1 | 1496 | 103 | ||
| GenBank mRNA | AY739287.1 | - | 1496 | 103 | ||
| BT085747.1 | - | 1401 | 103 | |||
| EU964296.1 | - | 1427 | 103 | |||
More Information
Set Membership: |
|

pdi4
Genomic Map