Assay Details
| Gene Symbol: PDIL5-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112290.1 | NP_001105760.1 | 160 | 63 | ||
| GenBank mRNA | AY739291.1 | - | 160 | 63 | ||
| BT065663.1 | - | 129 | 63 | |||
More Information
Set Membership: |
|

PDIL5-1
Genomic Map