Assay Details
| Gene Symbol: pdi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112293.1 | NP_001105763.1 | 1149 | 112 | ||
| GenBank mRNA | AY739295.1 | - | 1149 | 112 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdi5| Gene Symbol: pdi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112293.1 | NP_001105763.1 | 1149 | 112 | ||
| GenBank mRNA | AY739295.1 | - | 1149 | 112 | ||
Set Membership: |
|