Assay Details
| Gene Symbol: sgo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112321.1 | NP_001105791.1 | 435 | 72 | ||
| GenBank mRNA | AY964185.1 | - | 435 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sgo1| Gene Symbol: sgo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112321.1 | NP_001105791.1 | 435 | 72 | ||
| GenBank mRNA | AY964185.1 | - | 435 | 72 | ||
Set Membership: |
|