Assay Details
| Gene Symbol: AGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112324.1 | NP_001105794.1 | 2360 | 104 | ||
| GenBank mRNA | AF497511.2 | - | 2360 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGA2| Gene Symbol: AGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112324.1 | NP_001105794.1 | 2360 | 104 | ||
| GenBank mRNA | AF497511.2 | - | 2360 | 104 | ||
Set Membership: |
|