Assay Details
| Gene Symbol: si603015d01b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112344.1 | NP_001105814.1 | 1014 | 62 | ||
| GenBank mRNA | DQ436921.1 | - | 1014 | 62 | ||
More Information
Set Membership: |
|

si603015d01b
Genomic Map