Assay Details
Gene Symbol: lon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112433.1 | NP_001105903.1 | 1641 | 62 | ||
GenBank mRNA | AY109454.1 | - | 1641 | 62 | ||
U85494.1 | - | 1641 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
