Assay Details
| Gene Symbol: LOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112527.1 | NP_001105997.1 | 1116 | 77 | ||
| GenBank mRNA | DQ335770.1 | - | 1116 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOX12| Gene Symbol: LOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112527.1 | NP_001105997.1 | 1116 | 77 | ||
| GenBank mRNA | DQ335770.1 | - | 1116 | 77 | ||
Set Membership: |
|