Assay Details
| Gene Symbol: Mre11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112579.1 | NP_001106049.1 | 1648 | 85 | ||
| GenBank mRNA | EF599099.1 | - | 1648 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mre11B| Gene Symbol: Mre11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112579.1 | NP_001106049.1 | 1648 | 85 | ||
| GenBank mRNA | EF599099.1 | - | 1648 | 85 | ||