Assay Details
| Gene Symbol: hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112606.1 | NP_001106076.1 | 820 | 79 | ||
| GenBank mRNA | AF033496.1 | - | 820 | 79 | ||
| AY103669.1 | - | 822 | 79 | |||
| BT085924.1 | - | 815 | 79 | |||
More Information
Set Membership: |
|

hsbp1
Genomic Map