Assay Details
| Gene Symbol: LOC100191165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136600.1 | NP_001130072.1 | 950 | 72 | ||
| GenBank mRNA | BT033189.1 | - | 950 | 72 | ||
| EU965596.1 | - | 962 | 72 | |||
More Information
Set Membership: |
|

LOC100191165
Genomic Map