Assay Details
| Gene Symbol: BAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136610.1 | NP_001130082.1 | 654 | 92 | ||
| GenBank mRNA | BT033200.1 | - | 654 | 92 | ||
| BT061081.1 | - | 596 | 92 | |||
| HQ858764.1 | - | 282 | 92 | |||
| JX122765.1 | - | 282 | 92 | |||
| Gene Symbol: LOC100285252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158146.1 | NP_001151618.1 | 627 | 80 | ||
| GenBank mRNA | EU971416.1 | - | 627 | 80 | ||
More Information
Set Membership: |
|

BAD1
Genomic Map