Assay Details
| Gene Symbol: LOC100191280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136714.1 | NP_001130186.1 | 1225 | 75 | ||
| GenBank mRNA | BT033326.1 | - | 1225 | 75 | ||
| BT039228.1 | - | 910 | 75 | |||
More Information
Set Membership: |
|

LOC100191280
Genomic Map