Assay Details
| Gene Symbol: LOC100192597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137812.1 | NP_001131284.1 | 416 | 88 | ||
| GenBank mRNA | BT034619.1 | - | 399 | 88 | ||
| EU958368.1 | - | 416 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100192597| Gene Symbol: LOC100192597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137812.1 | NP_001131284.1 | 416 | 88 | ||
| GenBank mRNA | BT034619.1 | - | 399 | 88 | ||
| EU958368.1 | - | 416 | 88 | |||