Assay Details
| Gene Symbol: LOC100193293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138434.1 | NP_001131906.1 | 541 | 89 | ||
| GenBank mRNA | BT035517.1 | - | 541 | 89 | ||
More Information
Set Membership: |
|

LOC100193293
Genomic Map