Assay Details
| Gene Symbol: LOC100193477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138592.1 | NP_001132064.1 | 535 | 79 | ||
| GenBank mRNA | BT035745.1 | - | 365 | 79 | ||
| EU961370.1 | - | 535 | 79 | |||
More Information
Set Membership: |
|

LOC100193477
Genomic Map