Assay Details
| Gene Symbol: LOC100193689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138781.1 | NP_001132253.1 | 651 | 74 | ||
| GenBank mRNA | BT036021.1 | - | 651 | 74 | ||
More Information
Set Membership: |
|

LOC100193689
Genomic Map