Assay Details
| Gene Symbol: LOC100193739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138826.1 | NP_001132298.1 | 1174 | 98 | ||
| GenBank mRNA | BT036086.1 | - | 1174 | 98 | ||
More Information
Set Membership: |
|

LOC100193739
Genomic Map