Assay Details
| Gene Symbol: LOC100194380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139414.1 | NP_001132886.1 | 1329 | 66 | ||
| GenBank mRNA | BT036911.1 | - | 1329 | 66 | ||
More Information
Set Membership: |
|

LOC100194380
Genomic Map