Assay Details
| Gene Symbol: UBF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138130.1 | NP_001131602.1 | 556 | 70 | ||
| GenBank mRNA | BT035083.1 | - | 556 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBF9| Gene Symbol: UBF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138130.1 | NP_001131602.1 | 556 | 70 | ||
| GenBank mRNA | BT035083.1 | - | 556 | 70 | ||
Set Membership: |
|