Assay Details
| Gene Symbol: LOC100194325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139363.1 | NP_001132835.1 | 1397 | 61 | ||
| GenBank mRNA | BT036842.1 | - | 1397 | 61 | ||
| EU961417.1 | - | 1309 | 61 | |||
More Information
Set Membership: |
|

LOC100194325
Genomic Map