Assay Details
| Gene Symbol: LOC100217142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143508.1 | NP_001136980.1 | 700 | 71 | ||
| GenBank mRNA | BT038003.1 | - | 700 | 71 | ||
More Information
Set Membership: |
|

LOC100217142
Genomic Map