Assay Details
| Gene Symbol: srp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143562.1 | NP_001137034.1 | 364 | 71 | ||
| GenBank mRNA | BT036022.1 | - | 275 | 71 | ||
| BT038120.1 | - | 364 | 71 | |||
More Information
Set Membership: |
|

srp32
Genomic Map