Assay Details
| Gene Symbol: srp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143562.1 | NP_001137034.1 | 632 | 93 | ||
| GenBank mRNA | BT036022.1 | - | 543 | 93 | ||
| BT038120.1 | - | 632 | 93 | |||
More Information
Set Membership: |
|

srp32
Genomic Map